Topaz GC Inlet Liners for Thermo Gas Chromatographs, Restek
Supplier: Restek
23438 23325 23436 23437 23441 23445 23446 23323 23439 23448 23278
75877-866PK 494.48 CAD
75877-866 75877-892 75877-902 75877-904 75877-906 75877-912 75877-914 75878-212 75878-218 75878-256 75878-262
Topaz GC Inlet Liners for Thermo Gas Chromatographs, Restek
Chromatography Supplies Chromatography Inlet Liners
Topaz GC inlet liners feature revolutionary technology and inertness to deliver you the next level of True Blue Performance.
  • Topaz technology deactivation
  • Low breakdown
  • Reproducible
  • Maximized GC uptime
These inlet liners feature low breakdown for accurate and precise low-level GC analyses. Liners undergo manufacturing controls and QC testing for superior reliability across compound classes, and they feature unparalleled cleanliness for maximized GC uptime and lab throughput.
Certifications: Patented.