Topaz GC Inlet Liners for Shimadzu Gas Chromatographs, Restek
Supplier: Restek
23457 23321 23318 23279 23456 23471
75877-864PK 469.4 CAD
75877-864 75877-888 75878-240 75878-264 75878-302 75878-312
Topaz GC Inlet Liners for Shimadzu Gas Chromatographs, Restek
Chromatography Supplies Chromatography Inlet Liners
Topaz GC inlet liners feature revolutionary technology and inertness to deliver you the next level of True Blue Performance.
  • Topaz technology deactivation
  • Low breakdown
  • Reproducible
  • Maximized GC uptime
These inlet liners feature low breakdown for accurate and precise low-level GC analyses. Liners undergo manufacturing controls and QC testing for superior reliability across compound classes, and they feature unparalleled cleanliness for maximized GC uptime and lab throughput.