Topaz GC Inlet Liners for PerkinElmer Gas Chromatographs, Packed, Restek
Supplier: Restek
23451 23317 23449 23450 23452
75877-918PK 275.97 CAD
75877-918 75878-210 75878-230 75878-258 75878-296
Topaz GC Inlet Liners for PerkinElmer Gas Chromatographs, Packed, Restek
Chromatography Supplies Chromatography Inlet Liners
Topaz GC inlet liners feature revolutionary technology and inertness to deliver you the next level of True Blue Performance.
  • Topaz technology deactivation
  • Low breakdown
  • Reproducible
  • Maximized GC uptime
These inlet liners feature low breakdown for accurate and precise low-level GC analyses. Liners undergo manufacturing controls and QC testing for superior reliability across compound classes, and they feature unparalleled cleanliness for maximized GC uptime and lab throughput.
Certifications: Patented.
Order Now
SPECIFICATIONS
Environmentally Preferable