Topaz GC Inlet Liners for Bruker/Varian Gas Chromatographs, Restek
Supplier: Restek
23331 23463 23329 23465 23460 23461
75877-896PK 257.06 CAD
75877-896 75877-920 75878-252 75878-270 75878-272 75878-274
Topaz GC Inlet Liners for Bruker/Varian Gas Chromatographs, Restek
Chromatography Supplies Chromatography Inlet Liners
Topaz GC inlet liners feature revolutionary technology and inertness to deliver you the next level of True Blue Performance.
  • Topaz technology deactivation
  • Low breakdown
  • Reproducible
  • Maximized GC uptime
These inlet liners feature low breakdown for accurate and precise low-level GC analyses. Liners undergo manufacturing controls and QC testing for superior reliability across compound classes, and they feature unparalleled cleanliness for maximized GC uptime and lab throughput.
Certifications: Patented.