Screw Caps with Rubber Liners, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
73802U-8425 73802U-13425 73802U-15425 73802U-18400 73802U-20400 73802U-24400
66014-390CS 563.91 CAD
66014-390 66014-391 66014-392 66014-393 66014-394 89003-436
Screw Caps with Rubber Liners, Kimble Chase, DWK Life Sciences
Closures Bottle Closures
Urea screw thread caps with PTFE-faced foam-backed rubber liners
Ideal for use with scintillation vials.