Aluminium Heating Blocks, Kimble Chase, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
720200-0001 720200-0002 720205-0001 720210-0001
KT720200-0001EA 203.5 CAD
KT720200-0001 KT720200-0002 KT720205-0001 KT720210-0001
Aluminium Heating Blocks, Kimble Chase, DWK Life Sciences
Block Heaters Blocks
Ideal for use in place of a sand bath in microscale experiments.
  • Excellent heat transfer
  • Does not interfere with magnetic stirring