Four-Station Workbenches with Lockers
Supplier: Diversified Woodcrafts
WB12-0V WB12-4V
470226-024EA 10089.25 CAD
470226-024 470226-026
Four-Station Workbenches with Lockers
Furniture Tables and Workbenches
The base of this spacious workbench, Made in the USA, consists of a double-faced heavy gauge steel unit, welded and riveted throughout.
  • 20-gauge steel, all-welded, one-section construction with reinforced side panels
  • 12 vertical locker steel lockers with gray baked enamel finish
  • 18-gauge steel lockers have vented doors have spring loaded hinges, padlock hasp, and knockout plug for cylinder locks
  • Available with or without four vises
Lockers W×D×H: 12×21×15"
Top: 2¹/₄" maple
Limited Lifetime Warranty
Certifications: SEFA compliant and MAS Certified Green