Topaz GC Inlet Liners for Agilent Gas Chromatographs, Restek
Supplier: Restek
23434 23283 23433 23311 23313 23430 23428 23333 23306 23429 23431 23315 23282 23280 23301 23312 23308 23310 23302
75877-934PK 925.38 CAD
75877-934 75877-868 75877-932 75877-878 75877-884 75877-872 75877-874 75878-280 75878-254 75878-286 75877-858 75877-936 75878-268 75878-278 75878-266 75878-228 75878-216 75877-880 75878-208
Topaz GC Inlet Liners for Agilent Gas Chromatographs, Restek
Chromatography Supplies Chromatography Inlet Liners
Topaz GC inlet liners feature revolutionary technology and inertness to deliver you the next level of True Blue Performance.
  • Topaz technology deactivation
  • Low breakdown
  • Reproducible
  • Maximized GC uptime
These inlet liners feature low breakdown for accurate and precise low-level GC analyses. Liners undergo manufacturing controls and QC testing for superior reliability across compound classes, and they feature unparalleled cleanliness for maximized GC uptime and lab throughput.