Topaz GC Inlet Liners for DANI Gas Chromatographs, Restek
Supplier: Restek
23294 23292
75877-922PK 571.72 CAD
75877-922 75877-924 75877-926 75877-928 75878-220 75878-226
Topaz GC Inlet Liners for DANI Gas Chromatographs, Restek
Chromatography Supplies Chromatography Inlet Liners
Topaz GC inlet liners feature revolutionary technology and inertness to deliver you the next level of True Blue Performance.
  • Topaz technology deactivation
  • Low breakdown
  • Reproducible
  • Maximized GC uptime
These inlet liners feature low breakdown for accurate and precise low-level GC analyses. Liners undergo manufacturing controls and QC testing for superior reliability across compound classes, and they feature unparalleled cleanliness for maximized GC uptime and lab throughput.
Certifications: Patented.