Cogent Diol™ HPLC Column, MicroSolv Technology Corporation
Supplier: MicroSolv
Cogent Diol™
40260-02P-2 40060-HG5 40060-75P 40060-10P 40060-10P-3 40060-03P 40060-15P-3 40060-10P-1 40060-25P 40060-SP150 40060-03P-3 40060-05P 40060-03P-2 40060-HG4 40060-02P-3 40060-P21-150 40060-HG2 40060-03P-1 40060-05P-2 40060-02P-2 40060-HG1 40060-15P-2 40060-10P-2 40060-05P-3 40060-15P 40060-HG3 40060-05P-1 40060-HG6 40060-15P-1 40060-75P-2 40060-02P
10024-758EA 1590.24 CAD
10024-758 75839-648 75801-170 75839-662 75839-660 10024-780 10024-782 10024-760 10024-342 10024-344 10024-784 10024-346 10024-744 10024-348 10024-746 75801-168 10024-748 75839-658 75839-656 75839-654 75839-652 75839-650 10024-350 10024-750 10799-344 10024-774 10024-752 10024-776 10024-754 10024-756 10024-778
Cogent Diol™ HPLC Column, MicroSolv Technology Corporation
Chromatography Columns
These useful HPLC columns have a direct silicon-carbon point attachment to the silica-hydride surface.
  • RP and ANP
  • Unique selectivity for polar compounds useful for SFC
  • TYPE-C™ silica
This combination gives very stable, efficient and unique HPLC columns that operate in 3 different domains: Reversed Phase (RP), Normal Phase (NP) and Aqueous Normal Phase (ANP).
These HPLC columns are LC-MS compatible and are suitable for SFC.
A very powerful tool for every separations lab.