Maintenance Parts and Accessories for MLA Pipettors, VistaLab
Supplier: VistaLab
8066 9031 9035 1700 9032 9033 9030 9034 9075 9076 9077 9091 9092 9093 9094 9095
53514-060EA 53.34 CAD
53514-060 53514-122 53514-134 53514-142 53514-106 53514-136 53514-138 75813-812 75813-822 75813-872 75813-874 75813-876 75813-878 75813-880 75813-882 75813-884 75813-886 75814-384
Maintenance Parts and Accessories for MLA Pipettors, VistaLab
Pipettes Micropipettes
All components are compatible with MLA series pipettors, including MLA D-Tipper Pipettors, MLA Macro Volume Pipettors, MLA Precision Pipettors, MLA Digital Pipettors, and MLA Micro D-Tipper Pipettors.
Pipettor Calibration Kits may also be used with Ovation® Macro Single-Channel Electronic Pipettors, Ovation® ESC Electronic Single Channel Pipettors, Ovation® QS Digital Adjustable Single Channel Pipettors, Ovation® M Ergonomic Mechanical Pipettors, and Ovation® Fixed Volume Pipettors.