Alkaline Lysis Solution, Teknova
Supplier: TEKNOVA CA
A0301 A0313 A0311 A0314 A0304 A0303 A0302
CA100216-410EA 728.52 CAD
CA100216-410 CA100217-152 CA100217-480 CA100217-980 CA100219-096 CA100219-254 CA100220-172
Alkaline Lysis Solution, Teknova
Buffers Extraction Buffers
Alkaline lysis solution is used in the alkaline lysis method of plasmid DNA purification to produce a cleared lysate solution.