Dissolution Capsule Sinkers for Apparatus 2, Quality Lab Accessories
Supplier: Quality Lab Accessories
Agilent QLA Sotax
CAPWHT-XS CAPWHT-XLSS CAPWHT-04 CAPWHT-LG SNK010-VK MAGWHT-VKTOP BSK008-01LG CAPWHT-XLS CAPWHT-XL CUSBSK-ALF BSK008-01 WIRESK-VK CUSBSK-JPMAG SNK008-VK CAPWST-23 SNK080-VK CAPWST-31 CAPWHT-VK CAPWHT-4S CAPWST-15 CAPWHT-2S SNK040-VK CUSBSK-JP CAPWHT-02 CAPWHT-SV BSK008-JP CAPWHT-TR CAPWHT-PC CAPWST-19 CAPWST-18 MAGWHT-VK CAPWHT-S99
11010-212EA 465.26 CAD
11010-212 11010-156 93001-040 11010-158 11010-472 11010-152 11010-010 11010-154 11010-534 11010-138 75848-862 11010-470 11010-150 75849-182 11010-146 89049-234 89173-160 11010-148 89039-804 93001-074 11010-164 11010-142 11010-144 11010-008 75848-996 11010-160 89009-498 11010-140 11010-162 11010-382 89132-990 89132-970
Dissolution Capsule Sinkers for Apparatus 2, Quality Lab Accessories
Weights Sinkers
QLA offers the widest variety of Capsule Sinkers in the Dissolution Industry.
  • Made of high quality 316 stainless steel or PTFE coated over music wire
  • All sinkers are made in the USA
  • Custom sinkers are available upon request
Home-made sinkers tend not to be reproducible and so commercially available sinkers are quite popular
USP (711) states: " A small, loose piece of non-reactive material such as not more than a few turns of wire helix may be attached to dosage units that would otherwise float. Other validated sinker devices may be used."
The choice of sinker will depend on the tablet or capsule. The sinker should have minimal contact with the doasge form as this can affect the dissolution rate. The sinker should be heavy enough to sink the tablet/capsule to the bottom of the vessel. There are many varieties of sinkers including spiral, basket, pronged and spring style.