Dissolution Calibration Tools, Quality Lab Accessories
Supplier: Quality Lab Accessories
Agilent QLA QLA Innovative Product
LEVEL-UN TACHCLIP DEPSET-MI VIBMET-UN DEPSET-25A EUROINDC-M DPSPPD-VK DEPSET-SB CNTGAG-MI TACHOM-UN VALTOL-KT2 VALTOL-ASTM SPCR80 DEPSET-25 DEPSET-45A DEPSET-PK DDAPT-PLUS GNGDEP DEPGAG-VK VERTMET-MI COLLAR-75 DEPSET-45 THERMD-LC DPSPBK-VK ASTMBALL-25 WOBMET-MI
89173-172EA 886.32 CAD
89173-172 11010-478 89173-170 75849-230 11010-214 89173-110 11010-374 11010-516 11010-216 93001-042 11010-536 75848-884 75848-880 75848-882 75848-902 11010-168 93001-072 89173-162 75849-200 11010-226 11010-228 75848-974 89133-070 89173-106 75848-878 89132-950
Dissolution Calibration Tools, Quality Lab Accessories
Dissolution Apparatus
QLA manufactures Calibrations tools that have been designed for use on most dissolution baths.
  • Mechanical calibration parameters are easily verified using QLA's family of Calibration Tools
All are supplied with NIST traceable calibration certificates and user manuals. These calibration tools are serialized and come with a 1 year calibration certificate. Recalibration services are also offered by QLA.
Certifications: All calibration tools are serialized and come with a 1 year NIST traceable calibration certificate.