Cogent Bidentate C8™ HPLC Columns, MicroSolv Technology Corporation
Supplier: MicroSolv Technology Corp
40008-05P-2 40008-10P-2 40008-15P-2 4000825P 40008-05P-1 40008-10P 40008-10P-1 40008-15P 40008-15P-1 4000825P1 4000875P 40008-05P-3 4000815P3 40008-10P-3 40208-02P-2 40208-03P-2 40208-03P-3 40208-05P-2 40208-10P-2 40208-15P-2 40208-75P-2 40008-P21-150 40008-P21-250 40008-MP100 40008-MP150 40008-P21-50 40008-P21-75 40008-SP100 40008-SP150 40008-SP250 40008-SP250-3M 40008-SP50 40008-MP50 40008-P21-100 40008-03P-1 40008-75P-1 40008-P30-50 40008-P30-75 40008-MP250 40008-MP75 40008-P30-100 40008-P30-150 40008-P30-250 40008-SP75 40208-75P-1 40208-75P-3 40008-02P 40008-02P-2 40008-02P-3 40008-03P-2 40008-03P-3 40008-25P-3 40208-02P-3 40208-03P-1 40208-05P-1 40208-05P-3 40208-10P-1 40208-10P-3 40208-15P-1 40208-15P-3
97034-946EA 1482.51 CAD
97034-946 97034-952 97034-958 97034-940 97034-944 97034-948 97034-950 97034-954 97034-956 97034-960 97034-962 97035-080 97035-082 89220-018 10022-518 10022-520 10022-522 10022-524 10022-526 10022-528 10799-292 10799-294 10799-296 10799-298 10799-300 10799-302 10799-304 10818-580 10818-582 10818-584 10818-586 10818-588 10818-590 75801-088 75801-090 75801-112 75801-114 75801-124 75801-126 75801-128 75801-130 75801-132 75801-134 75801-136 75801-386 75801-388 75839-452 75839-454 75839-460 75839-462 75839-498 75839-500 75839-502 75839-504 75839-506 75839-508 75839-664 75839-666 75839-818 75839-820 75839-822 75839-826 75839-828 75839-830 75839-832 75839-864 75839-866 75839-868 75839-870 75839-872 75839-874 75839-876 75839-878 75856-518 75856-520 75856-540
Cogent Bidentate C8™ HPLC Columns, MicroSolv Technology Corporation
Chromatography Columns
These Cogent Bidentate C8™ phases are moderately hydrophobic and for that reason are a good starting column when scouting for compounds of unknown polarity. They will perform in Normal Phase (NP), Reversed Phase (RP) and Aqueous Normal Phase (ANP).
  • Unique and extremely stable
  • Made with silica hydride particles
  • Excellent selectivity with efficient peaks
These C8 columns are produced with two points of attachment to the particle via direct silicon-carbon bonds.
All Cogent TYPE-C™ HPLC columns are made with silica hydride particles.
This makes them very unique and extremely stable under even harsh HPLC conditions producing excellent selectivity with efficient peaks. The service life time of these columns far exceeds normal C8 column expectations. Also, the Bidentate C8 does not need to be end-capped due the nature of the silica hydride surface.