HPLC Columns, YMC Basic
Supplier: YMC America,Inc
BA99S03-0502WT BA99S03-0503WT BA99S03-0546WT BA99S03-1002WT BA99S03-1003WT BA99S03-1046WT BA99S03-1502WT BA99S03-1503WT BA99S03-1546WT BA99S03-2546WT BA99S03-H502WT BA99S03-L502WT BA99S05-0546WT BA99S05-1002WT BA99S05-1046WT BA99S05-1502WT BA99S05-1503WT BA99S05-1506WT BA99S05-1546WT BA99S05-2503WT BA99S05-2506WT BA99S05-2546WT BA99S03-01Q1GC BA99S03-02Q1GC BA99S03-0504WT BA99S05-0502WT BA99S05-0503WT BA99S05-0504WT
97029-504EA 1389.96 CAD
97029-504 97029-508 97029-514 97029-516 97029-518 97029-522 97029-524 97029-526 97029-528 97029-530 97029-538 97029-540 97029-562 97029-566 97029-570 97029-572 97029-574 97029-576 97029-578 97029-580 97029-582 97029-584 89216-526 89216-530 89216-532 89216-540 89216-542 89216-544
HPLC Columns, YMC Basic
Chromatography Columns
The basic columns are unique reversed-phase column designed for analysis of pharmaceuticals and basic compounds.
  • Provides high resolution, and excellent peak shape
  • Useful for separation of peptides
The neutral silica support used is prepared by an advanced synthetic process which yields a chromatographic material with proprietary pore size and neutral silanols which minimizes interactions with both basic and acidic compounds.
There are no special surface treatments or exotic endcapping procedures so that the columns provides the same general selectivity seen with conventional C18 and C8 columns without basic compounds tailing.