ZetexPlus 200 Extreme Temperature Mitts, Newtex Industries
Supplier: Newtex Industries
2100038 2100039 2100038
32885-304PR 256.1 CAD
32885-304 32885-326 75794-906
ZetexPlus 200 Extreme Temperature Mitts, Newtex Industries
Hot Hand Protectors
ZetexPlus 200 mitts are for high-temperature applications.
  • Flame-resistant
  • Operating Temperature: 815 °C (1500 °F)
  • Maximum Temperature: 1095 °C (2000 °F)
ZETEXPLUS* 200 Mitt, For high temperature applications up to 815 °C (1500 °F). Offers excellent resistance to flames, sparks, and molten splash, Non-asbestos, high-temperature replacement for high-silica and ceramic fabrics, High tensile strength, Vermiculite.