ZetexPlus 200 Extreme Temperature Gloves, Newtex Industries
Supplier: Newtex
2100011 2100014 2100012
75794-910CS 4963.87 CAD
75794-910 75794-952 75794-944
ZetexPlus 200 Extreme Temperature Gloves, Newtex Industries
Gloves
ZetexPlus 200 gloves are for high-temperature applications.
  • Flame-resistant
  • Operating temperature: 815 °C (1500 °F)
  • Maximum temperature: 1095 °C (2000 °F)
ZetexPlus 200 gloves are for high temperature applications up to 815 °C (1500 °F). Offers excellent resistance to flames, sparks, and molten splash. Non-asbestos, high-temperature replacement for high-silica and ceramic fabrics. High tensile strength, vermiculite.