Trichoderma viride Cellulase

Supplier: MilliporeSigma

219466-200KU 219466-100KU
CA80603-592EA 1661.79 CAD
CA80603-592 CA80603-594
Trichoderma viride Cellulase
Enzymes

From Trichoderma viride This enzyme shows a very high filter paper decomposing activity, as well as, appreciable xylanase and hemicellulase activity.


It is an effective enzyme to isolate intact cells and protoplasts from plant tissues.

Order Now

SPECIFICATIONS

Enzyme name Cellulase
Environmentally Preferable
Source
Species Trichoderma viride
CAS No. 9012-54-8
Form Yellowish white solid
Storage conditions 2...8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR