BioClean™ P-ZERO™ Sterile Polychloroprene Gloves, Ansell
Supplier: Ansell Canada
BPZS65 BPZS70 BPZS75 BPZS80 BPZS85 BPZS90 BPZS60 BPZS65 BPZS70 BPZS75 BPZS80 BPZS85 BPZS90
75877-384CS 1262.69 CAD
75877-384 75877-386 75877-390 75877-392 75877-394 75877-396 75877-398 CA75877-384 CA75877-386 CA75877-390 CA75877-392 CA75877-394 CA75877-396 CA75877-398
BioClean™ P-ZERO™ Sterile Polychloroprene Gloves, Ansell
Gloves
BioClean™ P-Zero™ Sterile Polychloroprene Gloves offer excellent sensitivity and cytotoxic drug protection.
  • Chemical resistant
  • Excellent tactility
  • Double-donnable
  • Textured for improved grip
  • Non-particulating EasyTear™ packaging
Providing exceptional grip, sensitivity, and cytotoxic protection, BioClean™ P-Zero™ Sterile Polychloroprene Gloves have excellent antistatic properties and offer both chemical protection and outstanding tactility.
Certifications: Manufactured in a facility holding ISO 9001: 2008 and 89/686/EEC Article 11b certifications.
More Product Information