Acura® 846 Positive Displacement Micropipette, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
W851295 W851299 W851303 W851307 W851311
10842-430CS 989.61 CAD
10842-430 10842-432 10842-434 10842-436 10842-438
Acura® 846 Positive Displacement Micropipette, DWK Life Sciences
Pipettes Micropipettes
Equipped with an with ETFE tipped plunger, these micropipettes are ideal for pipetting high density, viscous, foaming, or volatile liquids.