Disposable Culture Tubes, Simport Scientific
Supplier: Simport Scientific
T400-3 T400-7A T400-3A T400-4 T400-7 T400-10 T400-3B T400-3G T400-3O T400-3Y T400-3AB T400-3AG T400-3AO T400-3AY T400-4A T400-10A T400-3DS T400-3AAM
CA60830-021CS 188.7 CAD
CA60830-021 CA60830-023 CA60830-025 CA60830-046 CA60830-070 CA60830-082 CA60830-162 CA60830-164 CA60830-166 CA60830-168 CA60830-172 CA60830-174 CA60830-176 CA60830-178 CA60830-202 CA73960-363 CA89032-006 CA10029-180
Disposable Culture Tubes, Simport Scientific
Tubes Culture Tubes
These culture tubes are ideal for use in bacteriology, RIA, coagulation, and other routine laboratory procedures.
Polypropylene tubes are translucent and will withstand an RCF of over 3000×g during centrifugation.
Polystyrene tubes are transparent and will withstand centrifugation speeds up to 1400×g. Clear plastic guarantees no danger of glass activation during testing. They also tolerate aqueous solutions, mild bases, and weak acids, but not organic solvents, aromatic or chlorinated hydrocarbons. Polystyrene tubes cannot be autoclaved.
Precision molding with virgin thermoplasticsMore Product Information