Dissolution Transdermal Testing Apparatus 5 and Apparatus 6, Quality Lab Accessories
Supplier: QLA
APPFIVEHRCLP TRANDE-SC APPFIVEHRSCR APPFIVE-HR APPSIX-01 APPSIX-EX APP5S16-35 APP5S20-35 APP5S30-35 APP5S40-35 APPFIVE-V35 APPFIVE-V56 APPFIVE-V80 APPSIX-19LC SPNAP6-01 AP6CYL-DK APP5S120-56 APP5S16-56 APP5S16-80 APP5S20-56 APP5S40-30 APP5S40-56 APP5S40-80 APPFIVE-HRGLA APPFIVE-V30
89049-182EA 12.78 CAD
89049-182 89049-332 89080-272 93001-024 93001-028 93001-030 97002-336 97002-338 97002-340 97002-342 97002-344 97002-346 97002-348 97002-350 97002-394 75849-112 75849-114 75849-116 75849-118 75849-120 75849-122 75849-124 75849-126 75849-130 75849-132
Dissolution Transdermal Testing Apparatus 5 and Apparatus 6, Quality Lab Accessories
Dissolution Apparatus
QLA manufactures APP 5 and APP 6 accessories for the dissolution industry.
  • All accessories meet current USP and Machine Manufacturer's specifications
  • Made in the USA
QLA offers numerous industry standard devices for Transdermal testing
For USP APP 5, both Hanson-style and VanKel-style patch retainers are compatible with all brands of dissolution baths. The transdermal patch is placed either between a glass disk and an inert mesh screen, or directly onto a stainless-steel mesh assembly. It is placed at the bottom of the vessel, with the release sideMore Product Information