Shaker Flasks with Six Standard Baffles, Delong® Style Neck, Chemglass
Supplier: Chemglass
CLS-2048-04 CLS-2048-05 CLS-2048-06 CLS-2048-08 CLS-2048-09 CLS-2048-10
30623-250EA 132.72 CAD
30623-250 30623-252 30623-254 30623-256 30623-258 30623-260
Shaker Flasks with Six Standard Baffles, Delong® Style Neck, Chemglass
Flasks Erlenmeyer Flasks
Flask has 6 baffles, 3 baffles on the lower outside edge of the flask and 3 baffles on the bottom surface of the flask to maximize oxygen transfer to the culture medium when used on an orbital shaker.
  • Delong® style neck is made to fit stainless steel or plastic closures
  • Longer neck reduces the amount of splashing that would reach the closure
Ordering information: Closures are sold separately. Contact your Avantor representative for more information.
Order Now
SPECIFICATIONS
Environmentally Preferable