Dissolution Cannula Filters, Quality Lab Accessories
Supplier: QLA
FIL001-HR FIL004-HR FIL010-01-A FIL010-01 FIL010-DK FIL010-HR-A FIL010-HR FIL035-01-A FIL035-01 FIL035-HR FIL045-DK FIL070-01 FIL070-DK FIL070-HR FIL10S-DK FIL10S-HR FIL70S-HR FIL090-DK FILTIP-05 FILTIP-10 FILTIP-20 FILTIP-45 FIL045-01-A FIL070-EW-A FIL45S-DK FIL45S-DK-A FIL001-CA FIL001-CA-A FIL001-PT FIL005-PT FIL005-PT-A FIL010-CA FIL010-CA-A FIL010-PT FIL010-PT-A FIL010-VK FIL010-VK-A FIL020-01 FIL020-01-A FIL020-EW FIL020-EW-A FIL020-HR FIL020-HR-A FIL035-CA FIL035-CA-A FIL035-VK FIL035-VK-A FIL045-HR-A FIL070-VK FIL070-VK-A FIL20S-DK FIL20S-DK-A FIL45S-HR FIL45S-HR-A BSK020-DKC FIL001-PT-A FIL004-CA FIL004-PT FIL004-PT-A FIL035-PT-A FIL040-CA FIL040-CA-A FIL070-CA FIL070-CA-A FIL070-PT FIL070-PT-A FIL070-ST FIL070-ST-A FIL010-01K-A FIL010-ST-A FIL020-ST FIL035-ST-A FIL020-ST-A FIL20S-HR FIL001-EW-A FIL001-EW FIL004-EW-A FIL004-EW FIL010-EW-A FIL010-EW FIL035-EW FIL035-EW-A FIL040-EW-A FIL040-EW FIL045-01 FIL070-EW FIL10S-DK-A FIL35S-DK-A FIL35S-DK FIL35S-HR FIL35S-HR-A FIL70S-DK FIL70S-DK-A FILTIP-02 FIL10S-HR-A FIL001-HR-A FIL001-01 FIL001-01-A FIL002-01 FIL002-01-A FIL020-DK FIL020-DK-A FIL020-PT FIL020-PT-A FIL035-DK FIL035-DK-A FIL035-PT FIL045-DK-A FIL045-VK FIL045-VK-A FIL070-DK-A FIL090-DK-A FIL10S-ST FIL20S-HR-A
11010-252PK 1647.94 CAD
11010-252 11010-256 11010-258 11010-260 11010-262 11010-264 11010-266 11010-268 11010-270 11010-274 11010-276 11010-280 11010-282 11010-286 11010-288 11010-292 11010-296 11010-298 11010-304 11010-306 11010-308 11010-310 89039-816 89039-818 89039-820 89039-822 89049-240 89049-242 89049-244 89049-246 89049-248 89049-250 89049-252 89049-254 89049-256 89049-258 89049-260 89049-262 89049-264 89049-266 89049-268 89049-270 89049-272 89049-274 89049-276 89049-278 89049-280 89049-284 89049-286 89049-288 89049-290 89049-292 89049-294 89049-296 89080-280 89080-294 89080-296 89080-300 89080-302 89080-306 89080-308 89080-310 89080-312 89080-314 89080-316 89080-318 89080-320 89080-322 89132-998 89133-002 89133-004 89133-012 89173-112 89173-114 93000-984 93000-986 93000-988 93000-990 93000-992 93000-994 93000-996 93000-998 93001-000 93001-002 93001-004 93001-006 93001-010 93001-012 93001-014 93001-016 93001-018 93001-020 93001-022 11010-302 11010-290 11010-250 75848-904 75848-906 75848-908 75848-910 75848-914 75848-916 75848-918 75848-920 75848-922 75848-924 75848-926 75848-928 75848-930 75848-932 75848-934 75848-936 75848-938 75848-940
Dissolution Cannula Filters, Quality Lab Accessories
Filters Filter Tubes
Only QLA manufactures pure quality filters which are guaranteed chromium and heavy metal free.
  • Made of the purest and highest quality UHMW Polyethylene (and PVDF where applicable)
  • All filters are made in the USA
  • Available in 100 filter packs or 1000 filter jars
Filters for most sampling methods are available in numerous micron ratings and three main types: cannula filters, filter discs, and filter tips. The PureQuality certification program guarantees filters not to have any chromium, heavy metals, material contaminants, or inorganic elemental interference. In addition, they have been formulated to provide the specified micron rating and verified by porisometer testing. All filters are USP compliant where applicable, and meet machine manufacturer specifications.
Certifications: All filters are supplied with a Certificate of Conformance (COC) stating that they are USP compliant, where applicable, and that they meet machine manufacturer specifications.