Glas-Col® Mantle, Sperical Flask, Series TM, Chemglass
Supplier: Chemglass
CG-12000-02 CG-12000-05 CG-12000-07 CG-12000-08 CG-12000-09 CG-12000-10 CG-12000-11 CG-12000-12 CG-12000-13 CG-12000-14 CG-12000-15
80060-622EA 825.43 CAD
80060-622 80060-624 80060-626 80060-628 80060-630 80060-632 80060-634 80060-636 80060-638 80060-640 80060-642
Glas-Col® Mantle, Sperical Flask, Series TM, Chemglass
Heaters Heating Mantles
The Glas-Col® Series TM spherical flask mantle covers only the flask’s bottom half, letting you see its full contents
All mantles are 115 Volt with the exception of the 72L which is 230V and are supplied complete with a 4-foot, 3-wire cord and locking connector. The 50 and 72L sizes are furnished with a terminal box instead of a cord. Terminal box models are available with switches and pilot lights.
Larger sizes have multiple circuits for ease of temperature control when a flask is less than half full.