Chemical OSHA Danger Signs, Hazardous, National Marker
Supplier: ACCUFORM MANUFACTURING, INC
D140AP D140P D140PB D140R D140RB D164AB D164PB D164RB D441AB D441P D441PB D441R D441RB D442AB D442P D442PB D442R D442RB D548AB D548PB D548RB
94008-452PK 54.1 CAD
94008-452 94008-454 94008-456 94008-458 94008-460 94008-554 94008-556 94008-558 94009-944 94009-946 94009-948 94009-950 94009-952 94009-954 94009-956 94009-958 94009-960 94009-962 94009-964 94009-966 94010-774 94010-776 94010-778
Chemical OSHA Danger Signs, Hazardous, National Marker
Signs
OSHA signs are the perfect way to alert your employees and guests when a hazard is present in your facility.
  • Ideal for indoor and outdoor labeling
  • Available in Alum, PS vinyl, Fiber glass and Rigid plastic (thick polystyrene) materials
  • .040 alum is weatherproof, non-corrosive and has baked-on enamel on both sides
  • .004 PS vinyl material has pressure sensitive adhesive backing
  • .095 fiber glass material is rigid, shatter-proof and dent resistant
  • .050 rigid plastic material has radius corners and pre-drilled corner mounting holes
Keep your facility safe, buy an OSHA regulated sign today and prevent an accident tomorrow. Each sign header reads "Danger".
Certifications: Comply with OSHA 29 CFR 1910.