Human Recombinant IL-4 (from P. pastoris)
Supplier: IBI Scientific
RPH-121 RPH-122 RPH-123 RPH-120 RPH-124 RPH-125 RPH-126
10039-784EA 771.45 CAD
10039-784 10039-786 10039-788 10039-932 10039-934 10039-936 10039-938
Human Recombinant IL-4 (from P. pastoris)
Proteins and Peptides
Human protein IL-4 induces differentiation of naïve helper T-cells to TH-2 cells. Upon activation by IL-4, TH2 cells subsequently produce additional IL-4 cells