HISTOSETTE® II Cassettes in QuickLoad ™ Sleeve, Simport
Supplier: Simport Scientific
M485-2SL M485-3SL M485-4SL M485-6SL M485-7SL M485-8SL M485-9SL M485-11SL M485-12SL M485-16SL M485-17SL M485-18SL M486-2SL M486-3SL M486-4SL M486-5SL M486-6SL M486-8SL M486-9SL M486-10SL M486-11SL M486-12SL M486-16SL M486-17SL M486-18SL M485-5SL M485-10SL M486-7SL M485-8SL M485-9SL M485-10SL M486-9SL M486-10SL M486-11SL M486-12SL M485-11SL M485-12SL M486-2SL M486-3SL M486-4SL M486-5SL M485-2SL M485-3SL M485-4SL M485-5SL M485-6SL M485-7SL M486-6SL M486-7SL M486-8SL M485-16SL M485-17SL M485-18SL M486-16SL M486-17SL M486-18SL
CA87002-368CS 538.2 CAD
CA87002-368 CA87002-370 CA87002-372 CA87002-376 CA87002-378 CA87002-380 CA87002-382 CA87002-386 CA87002-388 CA87002-390 CA87002-392 CA87002-394 CA87002-396 CA87002-398 CA87002-400 CA87002-402 CA87002-404 CA87002-408 CA87002-410 CA87002-412 CA87002-414 CA87002-416 CA87002-418 CA87002-420 CA87002-422 CA87002-374 CA87002-384 CA87002-406 75878-428 75878-430 75878-432 75878-434 75878-436 75878-438 75878-440 75878-442 75878-444 75878-446 75878-448 75878-450 75878-452 75878-458 75878-460 75878-462 75878-464 75878-466 75878-468 75878-470 75878-472 75878-474 76299-252 76299-254 76299-256 76299-258 76299-260 76299-262
HISTOSETTE® II Cassettes in QuickLoad ™ Sleeve, Simport
Embedding Cassettes Biopsy Processing Cassettes
Save time and money with convenient sleeved cassettes.
  • Suitable for hoppers accepting plastic sleeves in labeling instruments
  • Cassettes are resistant to chemical action of histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper draingage
Specially designed cassettes are molded from a special high density polymer designed to keep specimens safely submerged in liquid.
Anterior printing area is a 45 degree angle, offering an unobstructed view of the writing surface and making the cassette perfectly suitable to beMore Product Information