DNA/RNA Oxidative Damage (Clone 7E6.9) ELISA Kit, Cayman Chemical Company
Supplier: Cayman Chemical Company
For the measurement of 8-hydroxy-2'-deoxyguanosine and 8-hydroxyguanosine.
- Measure DNA oxidative damage marker 8-hydroxy-2’-deoxyguanosine and RNA damage marker 8-hydroxyguanosine with equal selectivity and sensitivity
- Cross reactivity of clone 7E6.9 validated by mass spectrometry
- Assay 24 samples (urine or other matrices) in triplicate or 36 samples in duplicate
- Measure 8-hydroxyguanosine levels down to 0.45 ng/ml
- Incubation: 18 hours | Development: 90-120 minutes | Read: Colorimetric at 405-420 nm
Cayman's DNA/RNA Oxidative Damage (Clone 7E6.9) ELISA Kit is a competitive assay that can be used to measure both 8-hydroxy-2'-deoxyguanosine and 8-hydroxyguanosine. This kit has been validated for use in urine, but other sample types may be used. The assay has a range from 0.15-20 ng/ml and a sensitivity (80% B/B0) of approximately 0.45 ng/ml.
Delivery information: Kit contains: Antibody, tracer, standard, buffers, reagents, and plate(s).
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...