Prostaglandin D₂ (PGD₂) ELISA Kit, Cayman Chemical
Supplier: Cayman Chemical Company
A competitive assay that can be used for quantification of PGD₂.
- Measure PGD₂ in multiple sample types
- Assay 24 samples in triplicate or 36 samples in duplicate
- Measure PGD₂ levels down to 55 pg/ml
- Incubation: 18 hours | Development: 90-120 minutes | Read: Colorimetric at 405-420 nm
Prostaglandin D2 (PGD₂) is biosynthesized in the brain by a soluble, 26 kDa glutathione-independent lipocalin-type PGD₂ synthase. This PGD₂ accumulates in the cerebrospinal fluid (CSF), where it induces physiologic sleep in rats and humans. PGD₂ is also synthesized in mast cells and leukocytes by a cellular, myeloid-type, glutathione-dependent PGD synthase. This PGD₂ which is formed in the intracellular and vascular compartments is rapidly metabolized to 11β-PGF2α and other metabolites. Measurement of the parent eicosanoid PGD₂ is appropriate in cell culture lysates and in CSF, where concentrations of several hundred pg/ml have been measured. Cayman’s PGD₂ ELISA Kit is a competitive assay that can be used for quantification of PGD₂ in cell culture lysates and enzymatic reactions.
Delivery information: Kit contains: Antibody, Tracer, Standard, Buffers, Reagents, and Plate