Micro® Green Clean Biodegradable Cleaner, International Products Corporation
Supplier: International Products
M-1050-12 M-1052 M-1053-055
75854-300CS 973.23 CAD
75854-300 75854-302 75854-304
Micro® Green Clean Biodegradable Cleaner, International Products Corporation
Cleaning Agents Detergents and Surface Cleansers
Micro® Green Clean is an industrial-strength, free-rinsing, biodegradable, multi-purpose hard surface cleaner.
  • Typical pH of 9.5
  • Free-rinsing; does not leave interfering residue
  • Biodegradable and phosphate-free
  • NSF-registered as a USDA-A1 cleaner
  • Fast, effective soil removal
  • ISO-Certified Quality System
Micro® Green Clean is designed for use in a wide range of cleaning applications
Micro® Green Clean is registered with NSF as an A1 cleaner and can be validated in FDA processes. Validation methods are available upon request. Typical pH: 9.5.
Excellent for use on stainless steel, glass, ceramic, plastic, rubber, concrete, and most other hard surfaces.
Excellent for cleaning metals, ceramics, medical instruments, food-processing equipment, filter membranes, and other surfaces.
Accessories information: Accessories not included.