Micro® A07 Citric Acid Biodegradable Cleaner, International Products Corporation
Supplier: International Products
M-0801-12 M-0820 M-0890-055
75854-306CS 1052.63 CAD
75854-306 75854-308 75854-310
Micro® A07 Citric Acid Biodegradable Cleaner, International Products Corporation
Cleaning Agents Detergents and Surface Cleansers
Micro® A07 is a powerful blend of chelating citric acid and anionic surfactants.
  • Typical pH of 3.0
  • Free-rinsing; does not leave interfering residue
  • Biodegradable and phosphate-free
  • NSF-registered as a USDA-A1 cleaner
  • Fast, effective soil removal
  • ISO-Certified Quality System
Micro® A07 is designed to remove salts, soap scum, metal oxides, hard water scale, grease, rust, milkstone, mineral deposits, and inorganic material from filter membranes, labware, and industrial equipment
Registered with NSF as an A1 cleaner and can be validated in FDA processes. Validation methods are available upon request. Typical pH: 3.0.
Excellent for use on stainless steel, glass, ceramic, plastic, rubber, concrete, and most other hard surfaces.
Micro® A07 is non-corrosive and does not contain solvents, phosphates, silicates, phenols, or Substances of Very High Concern.
Accessories information: Accessories not included.