VWR® Flip Cap Centrifuge Tubes, Sterile
Supplier: VWR
VWR®
CFT221150 CFT284500 CFT221500 CFT211150 CFT211500
76404-664CS 791.26 CAD
76404-664 75845-208 75845-210 75845-214 75845-212
VWR® Flip Cap Centrifuge Tubes, Sterile
Tubes Centrifuge Tubes
Centrifuge tubes are ideal for use in centrifuging samples or storage in many research areas.
  • Manufactured from high-grade polypropylene
  • Easy flip-open caps enable single-handed access
  • Tubes feature black printed graduations and writing area can withstand chloroform
  • Autoclavable at 121 °C, and freezable to –80 °C
  • Packaged in zip sealed bags or environment friendly, recyclable paper racks
  • Gamma radiation sterilized, DNase-free, RNase-free, and non-pyrogenic
  • Leak-proof
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
Certifications: ISO 13485:2016 and ISO 9001:2015
Packaging: Packaged in zip sealed bags or environmental friendly, recyclable paper racks.