Flask Brushes, Justman Brush
Supplier: Justman Brush
J620368 J620370 J610372
17145-050EA 40.91 CAD
17145-050 17145-052 17145-054
Flask Brushes, Justman Brush
Brushes
These brushes with special construction puts the cleaning action directly where it is needed.
The bristles fold flat to fit throught the narrow bottle neck, then expand to fit the chamber.