MycoZap™ Prophylactic
Supplier: LONZA PHARMA - BIOSCIENCE CA
VZA-2031 VZA-2032
CA10064-510EA 298.23 CAD
CA10064-510 CA10064-512
MycoZap™ Prophylactic
MycoZap™ Prophylactic
MycoZap™ Antibiotics are extremely powerful combinations of innovative antibiotics for the protection of cell cultures from mycoplasma contamination. While MycoZap™ Prophylactic prevents mycoplasma contamination, MycoZap™ Plus offers complete protection against a broad range of common contaminants including mycoplasma.
Benefits:
Specifically prevents mycoplasma contamination
Also active against other species of the mycoplasma group like Acholeplasma and Spiroplasma
Applications:
Can be used in combination with other antibiotics (e.g., Pen/Strep) to prevent other microbial contaminants