MycoZap™ Mycoplasma Elimination Reagent, Lonza
Supplier: LONZA PHARMA - BIOSCIENCE CA
LT07-818 LT07-918
CA10064-176EA 458.2 CAD
CA10064-176 CA10064-178
MycoZap™ Mycoplasma Elimination Reagent, Lonza
Microbiology Test Systems Microorganism Tests
The MycoZap™ Mycoplasma Elimination Reagent allows efficient mycoplasma removal with minimal impact on cell viability.
  • Efficient mollicute elimination by a combination of antibiotic and antimetabolic agents
  • Eradicates Mycoplasma, Acholeplasma, Spiroplasma, and Entomoplasma
  • Minimal toxic effects
  • Suited for a broad range of cell cultures
The MycoZap™ Reagent can eliminate detectable mycoplasma contamination in as few as 4 days and has been optimized to clear mycoplasma with minimal toxic effects on the infected cells. It eliminates mycoplasma by using a combination of antibiotic and antimetabolic agents. This approach allows for a highly reliable elimination of  mycoplasma that cannot be achieved by the use of antibiotics alone. The MycoZap™ Reagent can be used to eradicate mollicutes, including Mycoplasma, Acholeplasma, Spiroplasma and Entomoplasma species in cell cultures.