Guanidinium thiocyanate ≥99% (by titrimetric analysis), Calbiochem®
Supplier: MilliporeSigma
368975-100GM
368975-500GM
CA80058-164EA
256.23
CAD
CA80058-164
CA80058-168
Guanidinium thiocyanate ≥99% (by titrimetric analysis), Calbiochem®
Guanidinium thiocyanate
Powerful protein denaturant widely used in purification of proteins and nucleic acids.
Formula:
CH₅N₃·CHNS Storage Temperature: Ambient |
MDL Number:
MFCD00013027 CAS Number: 593-84-0 |
Specification Test Results
Appearance | White solid |
Absorbance (1.7 M, H₂O, 280 nm) |
|
Purity (by titration) | ≥99% |
Toxicity | Harmful |
Storage | Store at 15 to 30 °C. Protect from moisture. |
Solubility | Ethanol (5%) or H₂O (25%) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...