AMCO CLEAR® Turbidity Standard, GFS Chemicals
Supplier: GFS CHEMICALS INC
85336
86698
86788
86789
85725
85726
85004
85006
71002-436EA
192.78
CAD
71002-436
71002-438
71002-440
71002-442
71002-444
71002-446
71002-448
71002-450
71002-452
AMCO CLEAR® Turbidity Standard, GFS Chemicals
Standards
Turbidity Standards
AMCO Clear® is an EPA approved primary standard for calibration of turbidity sensors.
- Safe, non-toxic and disposable
- easy-to-use with no dilution or resuspension
NIST traceable to particle size/distribution and optical density, and its inherent stability means it does not settle from suspension.
Certifications: Approved by the US EPA for drinking water analysis for turbidity.