Lance Blades, Cincinnati Surgical
Supplier: CINCINNATI SURGICAL CO.
92010 92011 92012 92015T 92020 92021 92022 92110 92111 92112 92115T 92120 92121 92122
75870-554BX 136.46 CAD
75870-554 75870-556 75870-558 75870-560 75870-562 75870-564 75870-566 75870-608 75870-610 75870-612 75870-614 75870-616 75870-618 75870-620
Lance Blades, Cincinnati Surgical
Scalpels and Scalpel Assemblies Scalpel Blades
Lance products enjoy a reputation of long lasting cutting edges
  • 100% visual inspection
  • Wide grinding angle resulting in high durability
  • Low surface finish
  • Limited range
Processed from British “cold rolled” steel which is superior in tolerance, concentricity, and straightness, allowing for wider grinding angles.
Certifications: CE marked.
Order Now
SPECIFICATIONS
Environmentally Preferable
Environmental certification ISO 14001 and ISO 50001