EZ Grip Tissue Culture Nosepiece, Drummond
Supplier: Drummond
4-000-079-W 4-000-087
80078-970EA 47.1 CAD
80078-970 89082-452
EZ Grip Tissue Culture Nosepiece, Drummond
Pipettes Pipette Controllers
Drummond EZ grip tissue culture nosepiece eliminates the possibility of cross contamination due to over pipetting.
  • Self-locking action of hydrophilic layer prevents further pipetting with contaminated filter
  • Double-membrane filter
It incorporates a dual-hydrophilic 0.8 μm, hydrophobic 0.8 μm filter, which prevents further use once contaminated by fluid.
Each kit contains all necessary parts including four filters.
This double membrane filter provides a barrier to particles as small as 0.1 μm.
Accessories information: Extra filters may beMore Product Information