PCR Films and Foils
Supplier: Eppendorf
0030132947 0030127854 0030127838 0030127781 0030127790 0030127838 0030127854 0030132947
CA89094-566CS 343.98 CAD
CA89094-566 CA89167-286 CA89167-284 75875-054 75875-056 75875-058 75875-060 75875-068
PCR Films and Foils
Seals Microplate Seals
Ensure airtight sealing of your PCR plates with Eppendorf® PCR films and foils. Tailor your choice to your assay needs, like the Masterclear® real-time PCR film, optimal for qPCR with its superior optical characteristics, or heat-sealing options for the best evaportion protection.
  • PCR plate seals offer excellent chemical resistance and have been tested for resistance to a variety of common laboratory substances
  • Specialized seals with optimal optical properties are available to help minimize signal loss in qPCR
Eppendorf PCR films and foils provide effective and airtight sealing for plates to help protect PCR samples during thermal cycling and storage.