DNA Fingerprinting Kit: Reunite the Siblings
Supplier: GENOSENSOR CORPORATION
4003 4004
470225-418EA 294.2 CAD
470225-418 470225-420
DNA Fingerprinting Kit: Reunite the Siblings
Educational Materials Biology Educational Materials PCR Learning Activities
Your students are asked to help a person determine which of three candidates is his long-lost relative.
  • Easy to follow protocols with setup and step-by-step guidance
  • Procedures contain questions allowing better assessment for understanding
  • Allows real life application of common scientific techniques
  • No human body fluids are handled in these experiments
A client has been searching for a sibling that has been accidentally separated from his family. He has identified three potential candidates and needs help determining which is the true sibling. Each candidate has submitted a DNA sample, as has the client. Student investigators are asked to determine which is the true sibling.
Kit I: Allows students to perform a restriction digest of four DNA samples and compare their sizes via Agarose Gel Electrophoresis.
Kit II: Allows students to amplify four DNA samples via Polymerase Chane Reaction (PCR) and observe visible bands via Agarose Gel Electrophoresis. Includes Master mix (including primers and enzyme) and DNA ladder.
The kit includes four samples of DNA (plasmids). One representing a man looking for his family, and three representing possible matches. After completing the experiment, students will not only solve the crime, but also cement their understanding of the genetic concepts associated with these two experiments.
Ordering information: Kits come with directions to download instructional material and procedures.
Equipment needed but not included: Heat block/water bath for restriction. Thermal cycler for PCR, microcentrifuge; vortex for sample preparation. Electrophoresis equipment/reagents including transilluminator/light box for analysis.
Delivery information: Kits include perishable materials, please provide a delivery date at check-out.
Packaging: Kits contain reagents only.