DNA Fingerprinting Kit: Identify the Criminal
Supplier: GENOSENSOR CORPORATION
4001 4002
470225-414EA 294.2 CAD
470225-414 470225-416
DNA Fingerprinting Kit: Identify the Criminal
Educational Materials Biology Educational Materials PCR Learning Activities
Your students are forensics professionals analyzing DNA collected at a crime scene.
  • Mock crime scene scenario designed to provide a fun and engaging learning experience
  • Procedures contain questions allowing better assessment for understanding
  • Easy to follow protocols with setup and step-by-step guidance
  • Allows real life application of common scientific techniques
  • No human body fluids are handled in these experiments
A DNA sample of unknown origin was collected at a crime scene. Your student-investigators have obtained 3 different DNA samples from their suspects and now must identify the criminal.
Kit I: Allows students to perform a restriction digest of four DNA samples and compare their sizes via Agarose Gel Electrophoresis.
Kit II: Allows students to amplify four DNA samples via Polymerase Chane Reaction (PCR) and observe visible bands via Agarose Gel Electrophoresis. Includes Master mix (including primers and enzyme) and DNA ladder.
The kit includes four samples of DNA (plasmids). Three representing suspects, and a fourth, "unknown" sample recovered from a crime scene. After completing the experiment, students will not only solve the crime, but also cement their understanding of the genetic concepts associated with these two experiments.
Ordering information: Kits come with directions to download instructional material and procedures.
Equipment needed but not included: Heat block/water bath for restriction. Thermal cycler for PCR, microcentrifuge; vortex for sample preparation. Electrophoresis equipment/reagents including transilluminator/light box for analysis.
Delivery information: Kits include perishable materials, please provide a delivery date at check-out.
Packaging: Kits contain reagents only.