What's the Source of the Food Poisoning?
Supplier: GENOSENSOR CORPORATION
3201 3202
470225-410EA 294.2 CAD
470225-410 470225-412
What's the Source of the Food Poisoning?
Educational Materials Biology Educational Materials PCR Learning Activities
Your students are medical professionals trying to save a patient's life by identifying a food-borne illness-causing microbe.
  • Easy to follow prodotocls with setup and step-by-step guidance
  • Procedures contain questions allowing better assessment for understanding
  • Real life application of commonly used scientific techniques
  • No human body fluids or bacteria are handled in these experiments
Imagine a patient came into the emergency room and was diagnosed with food poisoning and the doctors want to know which illness-causing microbe is responsible.
Kit I: Allows students to perform a restriction digest of four DNA plasmids and compare their sizes via Agarose Gel Electrophoresis.
Kit II: Allows students to amplify four DNA samples via Polymerase Chane Reaction (PCR) and observe visible bands via Agarose Gel Electrophoresis. Invludes Master mix (including primers and enzyme) and DNA ladder.
The kit includes four DNA plasmids. Three representing a microbe, and a fourth, "unknown" sample from the patient. After completing the experiment, students will not only discover which microbe is responsible, but they will also cement their understanding of the genetic concepts associated with these two experiments.
Ordering information: Kits come with directions to download instructional material and procedures.
Equipment needed but not included: Heat block/water bath for restriction. Thermal cycler for PCR, microcentrifuge; vortex for sample preparation. Electrophoresis equipment/reagents including transilluminator/light box for analysis.
Delivery information: Kits include perishable materials, please provide a delivery date at check-out.
Packaging: Kits contain reagents only.