Laboratory Dry Sterilizer, Yamato
Supplier: Yamato Scientific
SK-401-115V SK-401-220V SK-601-115V SK-601-220V
76550-280EA 6393.06 CAD
76550-280 76550-282 76550-284 76550-286
Laboratory Dry Sterilizer, Yamato
Sterilizers Autoclaves (Steam Sterilizers)
The low-cost, programmable laboratory dry sterilizer is easy to operate.
  • Two 30 mm dia. exhaust ports on top
  • Maximum temperature reaching time: ~70 min
  • Two shelf plates and four brackets included
  • Natural convection
  • Iron chromium wire heater
The sterilizer features a stainless steel interior and an exterior with a chrome-free, electro-galvanized carbon steel sheet coated with a chemical-proof, baked-on finish.
A green LED display indicates the measured temperature, and a red LED display indicates the setting temperature. An overheat prevention device and an earth leakage breaker are provided. Self-diagnostic functions ensure safe operation.
It's equipped with calibration offset, a lock function, and power-failure recovery mode. The natural convection oven has a temperature accuracy of 20 °C (at 260 °C).
Caution: High Temperature Components. Never use explosive or flammable material. Never use in flammable or explosive gas atmosphere.