Laboratory Stirrer, Yamato
Supplier: Yamato Scientific
LT-500AA LT-500BA LT-400AA LT-400BA LT-400CA
75846-416KT 3525.36 CAD
75846-416 75846-418 75846-652 75846-654 75846-656
Laboratory Stirrer, Yamato
Stirrers Overhead Stirrers
These laboratory stirrers feature a sensitive feedback system that keeps the set speed even with changing viscosity during stir.
  • Feedback Control
  • Panel Display: Digital Speed Display, Overload Display, Torque Indicator (20% Gradation)
  • DC Brushless Motor
  • Power Supply Cord with Bipolar Grounding-Type Plug: 2 m
  • Safety-Oriented Design
They also feature a maintenance-free DC brushless motor and a digital speed indicator for accurate speed setting and confirmation
The noise-prevention measures and safety-oriented design allow for an optimal workMore Product Information