Inert Oven, Yamato

Supplier: Yamato Scientific
DN-411IE DN-611IE
75846-556EA 15942.36 CAD
75846-556 75846-558
Inert Oven, Yamato
Ovens High Temperature Ovens
The Inert Oven is suitable for temperature tests and heat treatment in a non-oxidizing environment by introducing N2 gas into the chamber.

  • Heating Time: 60 Minutes
  • Shelf Support Quantity/Pitch: 12 Pieces/30 mm
  • Heater Circuit Control: TRIAC Zero-Crossing Method
  • Gradient: 12 °C (at 360 °C)

The oven's interior is made of stainless steel, and the exterior is made of a cold-steel plate with melamine-resin baking finish

It also comes equipped with various operation modes such as calibration offset, power failure recovery mode selection, and user configuration information saving. The digital PID controller supports fixed temperature, quick auto stop, auto stop and start, and program operations. Self-diagnostic features, a key lock function, and independent overheating prevention are provided. A green LED digital display shows the temperature, and an orange LED display shows the setting temperature. A monitor also displays power consumption, CO2 discharge, and heater operation.

It features heat treatment of up to 360 °C. It includes a condenser-type Sirocco fan, two shelves, and four brackets.

Caution: High-temperature components. Never use in flammable or explosive gas atmosphere. Never use explosive or flammable material.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR