VWR® Desiccator, Bottom and Lid with Glass Knob
Supplier: VWR International
75871-658 75871-660 75871-662 75871-664 75871-666
75871-658CS 122.26 CAD
75871-658 75871-660 75871-662 75871-664 75871-666
VWR® Desiccator, Bottom and Lid with Glass Knob
Desiccators Desiccators, Glass
This VWR® Desiccator is made from high quality Borosilicate 33 expansion glass
  • Very good chemical resistance
  • High thermal shock and pressure resistance, working with vacuum
  • 2 mm tolerance
  • Autoclavable
It is designed for a variety of vacuum applications, including desiccation of samples and storage of materials sensitive to humidity.