Quartz Low Form Beakers, Technical Glass Products
Supplier: Technical Glass Products
FQBLF10 FQBLF25 FQBLF50 FQBLF100 FQBLF250 FQBLF400 FQBLF600 FQBLF1000
75875-516EA 105.28 CAD
75875-516 75875-518 75875-520 75875-522 75875-524 75875-526 75875-528 75875-530
Quartz Low Form Beakers, Technical Glass Products
Beakers
Fused Quartz consists of high-purity Silica (SiO2) with a purity level of 99.998%
Fused Quartz also offers a number of unique properties that make it ideal in a variety of applications where other materials prove to be problematic. These beakers provide extremely high purity, a high softening temperature (1670°C), optical transparency over a wide range, low thermal expansion, excellent thermal shock, and resistance to a large number of elements and compounds.
Low Form Beakers manufactured from this material are invaluable in research and industry applications.